Product Details

SNP ID
rs9376684
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:142307998 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAATTTCATTTCTGGCCCAGGTGAT[C/T]CCAGAAGTGACCACAAAGTCAATTG
Phenotype
MIM: 612243
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ADGRG6 PubMed Links

Gene Details

Gene
ADGRG6
Gene Name
adhesion G protein-coupled receptor G6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001032394.2 Intron NP_001027566.1
NM_001032395.2 Intron NP_001027567.1
NM_020455.5 Intron NP_065188.4
NM_198569.2 Intron NP_940971.1
XM_005267061.3 Intron XP_005267118.1
XM_006715516.2 Intron XP_006715579.1
XM_006715517.2 Intron XP_006715580.1
XM_006715518.2 Intron XP_006715581.1
XM_011535964.1 Intron XP_011534266.1
XM_017011085.1 Intron XP_016866574.1

View Full Product Details