Product Details

SNP ID
hCV30198082
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:33165235 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATCTTTCTTATTTGCCCAGAATTC[C/T]TGAGTAGGCAGTTCAAGTGTGAGAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BPIFA2 PubMed Links

Gene Details

Gene
BPIFA2
Gene Name
BPI fold containing family A member 2
There are no transcripts associated with this gene.

Gene
LOC105372714
Gene Name
uncharacterized LOC105372714
There are no transcripts associated with this gene.

View Full Product Details