Product Details

SNP ID
rs9933310
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:29679583 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CGCTGGTAGCTCGGGAGAAGCAGAG[A/G]CAAAGTCCTGTCCCTTCTGCAGCTG
Phenotype
MIM: 606248 MIM: 182160
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
QPRT PubMed Links

Gene Details

Gene
QPRT
Gene Name
quinolinate phosphoribosyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318249.1 Intron NP_001305178.1
NM_001318250.1 Intron NP_001305179.1
NM_014298.4 Intron NP_055113.2
XM_005255223.3 Intron XP_005255280.2
XM_017023101.1 Intron XP_016878590.1
Gene
SPN
Gene Name
sialophorin
There are no transcripts associated with this gene.

View Full Product Details