Product Details

SNP ID
rs10417416
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:44651843 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCATATAGGATTTTAAATTCTCCAG[C/T]GGCCACCCCATTAAAAAATTAAAAG
Phenotype
MIM: 173850
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MIR4531 PubMed Links

Gene Details

Gene
MIR4531
Gene Name
microRNA 4531
There are no transcripts associated with this gene.

Gene
PVR
Gene Name
poliovirus receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135768.2 Intron NP_001129240.1
NM_001135769.2 Intron NP_001129241.1
NM_001135770.3 Intron NP_001129242.2
NM_006505.4 Intron NP_006496.4

View Full Product Details