Product Details

SNP ID
rs9713958
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:105375675 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGAAGAACTATTTCAGTATGAGCA[C/T]GAATTCCTTTTGCAGCAAATTGTAC
Phenotype
MIM: 601662
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ALCAM PubMed Links

Gene Details

Gene
ALCAM
Gene Name
activated leukocyte cell adhesion molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243280.1 Intron NP_001230209.1
NM_001243281.1 Intron NP_001230210.1
NM_001243283.1 Intron NP_001230212.1
NM_001627.3 Intron NP_001618.2

View Full Product Details