Product Details

SNP ID
rs9907259
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:35548265 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGTAGGCTGCCCTCTTTTCATAAA[A/G]CAGGTAGAGGTGTTTAATGGCTCTT
Phenotype
MIM: 614958
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLFN14 PubMed Links

Gene Details

Gene
SLFN14
Gene Name
schlafen family member 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001129820.1 2833 Missense Mutation CTT,TTT L905F NP_001123292.1
XM_017024576.1 2833 Missense Mutation CTT,TTT L905F XP_016880065.1
XM_017024577.1 2833 Missense Mutation CTT,TTT L905F XP_016880066.1
XM_017024578.1 2833 Missense Mutation CTT,TTT L905F XP_016880067.1
XM_017024579.1 2833 Missense Mutation CTT,TTT L905F XP_016880068.1

View Full Product Details