Product Details

SNP ID
rs9984726
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:44654285 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATAGGAGGCCACCTGCTCAGCAGCC[A/G]GTGGGGGTGCTCCAGGTGACAGGTC
Phenotype
MIM: 612920
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KRTAP10-11 PubMed Links

Gene Details

Gene
KRTAP10-11
Gene Name
keratin associated protein 10-11
There are no transcripts associated with this gene.

Gene
KRTAP12-3
Gene Name
keratin associated protein 12-3
There are no transcripts associated with this gene.

Gene
KRTAP12-4
Gene Name
keratin associated protein 12-4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198698.1 375 Silent Mutation ACC,ACT T110T NP_941971.1
Gene
TSPEAR
Gene Name
thrombospondin type laminin G domain and EAR repeats
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001272037.1 375 Intron NP_001258966.1
NM_144991.2 375 Intron NP_659428.2

View Full Product Details