Product Details

SNP ID
rs6685126
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:9159058 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGAAGCTGGTGGCTAAGAGGGGCT[C/T]CAGAGACTCCATCCAGATCTGCAGC
Phenotype
MIM: 611172
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MIR34A PubMed Links

Gene Details

Gene
MIR34A
Gene Name
microRNA 34a
There are no transcripts associated with this gene.

Gene
MIR34AHG
Gene Name
MIR34A host gene
There are no transcripts associated with this gene.

View Full Product Details