Product Details

SNP ID
rs10428360
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:2234086 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAAAGAAGACTATCAAAATACCCA[C/G]TAACAATAACCAGCTACAAAATTGA
Phenotype
MIM: 613430 MIM: 610887
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
HAUS3 PubMed Links

Gene Details

Gene
HAUS3
Gene Name
HAUS augmin like complex subunit 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303143.1 Intron NP_001290072.1
NM_024511.6 Intron NP_078787.2
Gene
POLN
Gene Name
DNA polymerase nu
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181808.3 Intron NP_861524.2

View Full Product Details