Product Details

SNP ID
rs9291318
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.4:47935710 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAATAGCCACTGCTGTCAGGTCTTA[A/G]TAATCAGAAAAAAATTAAAATTTAA
Phenotype
MIM: 123825
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
CNGA1 PubMed Links
Additional Information
For this assay, SNP(s) [rs76563043] are located under a probe and SNP(s) [rs138387210] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
CNGA1
Gene Name
cyclic nucleotide gated channel alpha 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000087.3 3022 Intron NP_000078.2
NM_001142564.1 3022 Intron NP_001136036.1
XM_005248049.4 3022 UTR 3 XP_005248106.2
XM_011513623.2 3022 Intron XP_011511925.1
XM_017007712.1 3022 Intron XP_016863201.1
Gene
LOC101927157
Gene Name
uncharacterized LOC101927157
There are no transcripts associated with this gene.

View Full Product Details