Product Details

SNP ID
rs9579637
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:30714904 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCGTTCCTGGCACTTATCATATTAC[C/G]CACTGTACTCCCCGCTTAGAGCTGT
Phenotype
MIM: 603700
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ALOX5AP PubMed Links

Gene Details

Gene
ALOX5AP
Gene Name
arachidonate 5-lipoxygenase activating protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204406.1 Intron NP_001191335.1
NM_001629.3 Intron NP_001620.2
XM_017020522.1 Intron XP_016876011.1

View Full Product Details