Product Details

SNP ID
rs10424540
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:985900 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCAGAAGATCATGTGCCCCGGGCC[A/T]GTCACCTGTCTGACTGCATCACCCA
Phenotype
MIM: 603265
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ARID3A PubMed Links

Gene Details

Gene
ARID3A
Gene Name
AT-rich interaction domain 3A
There are no transcripts associated with this gene.

Gene
WDR18
Gene Name
WD repeat domain 18
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024100.3 798 Silent Mutation CCA,CCT P82P NP_077005.2
XM_017027027.1 798 Silent Mutation CCA,CCT P94P XP_016882516.1

View Full Product Details