Product Details
- SNP ID
-
rs10251155
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:97989224 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGGATTACAGGTGTGAGCACCACAC[C/G]TGGCCCTTCAGTGATTTCTTGCTTT
- Phenotype
-
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
OCM2
PubMed Links
Gene Details
- Gene
- OCM2
- Gene Name
- oncomodulin 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_006188.3 |
|
Intron |
|
|
NP_006179.2 |
View Full Product Details