Product Details

SNP ID
rs10049681
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:94763229 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGAAGTGAGGCAGAATTCCTCTAAT[C/T]AGGTCTGAGTGTCATTTTCCTCTAA
Phenotype
MIM: 603248
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BMPR1B PubMed Links

Gene Details

Gene
BMPR1B
Gene Name
bone morphogenetic protein receptor type 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001203.2 Intron NP_001194.1
NM_001256792.1 Intron NP_001243721.1
NM_001256793.1 Intron NP_001243722.1
NM_001256794.1 Intron NP_001243723.1
XM_011532201.2 Intron XP_011530503.1
XM_017008558.1 Intron XP_016864047.1
XM_017008559.1 Intron XP_016864048.1
XM_017008560.1 Intron XP_016864049.1
XM_017008561.1 Intron XP_016864050.1
Gene
BMPR1B-AS1
Gene Name
BMPR1B antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

View Full Product Details