Product Details

SNP ID
rs10401542
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:43511761 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGATTTAAATGCATGAAAAATAAA[A/T]CCGCAAAGCTGTGACACACAACAAC
Phenotype
MIM: 608451
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ETHE1 PubMed Links

Gene Details

Gene
ETHE1
Gene Name
ETHE1, persulfide dioxygenase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320867.1 Intron NP_001307796.1
NM_001320868.1 Intron NP_001307797.1
NM_001320869.1 Intron NP_001307798.1
NM_014297.4 Intron NP_055112.2
XM_005258687.3 Intron XP_005258744.1
Gene
PHLDB3
Gene Name
pleckstrin homology like domain family B member 3
There are no transcripts associated with this gene.

View Full Product Details