Product Details
- SNP ID
-
rs5968237
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:83869794 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTTTTAGAAATAAAGGATAGATTAC[A/T]TAAAAAAATAGTTATTTTTATTTAT
- Phenotype
-
MIM: 300768
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
CYLC1
PubMed Links
Gene Details
- Gene
- CYLC1
- Gene Name
- cylicin 1
View Full Product Details