Product Details

SNP ID
rs9380953
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:40379018 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGATCTGGGGTCAGAAGTTCCCCTT[A/G]TCAAGTCCCAAGTCTTCCATCTAAG
Phenotype
MIM: 615676
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TDRG1 PubMed Links

Gene Details

Gene
TDRG1
Gene Name
testis development related 1 (non-protein coding)
There are no transcripts associated with this gene.

View Full Product Details