Product Details

SNP ID
rs10160292
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:44574416 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACCCACCCACCCTTGGCTTCCCCAT[G/T]TTGATCCTGATCTTGCTTGTTGGAA
Phenotype
MIM: 600623
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CD82 PubMed Links

Gene Details

Gene
CD82
Gene Name
CD82 molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001024844.1 Intron NP_001020015.1
NM_002231.3 Intron NP_002222.1
XM_006718223.2 Intron XP_006718286.1
XM_011520067.2 Intron XP_011518369.1

View Full Product Details