Product Details

SNP ID
rs9401094
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:118896510 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCAGCCATTCATAAAGCCAAAACGT[C/T]CCATTTTTAACTGGGATCCATTCAG
Phenotype
MIM: 609189 MIM: 610098
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ASF1A PubMed Links

Gene Details

Gene
ASF1A
Gene Name
anti-silencing function 1A histone chaperone
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014034.2 Intron NP_054753.1
Gene
MCM9
Gene Name
minichromosome maintenance 9 homologous recombination repair factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017696.2 Intron NP_060166.2
NM_153255.4 Intron NP_694987.1

View Full Product Details