Product Details

SNP ID
rs9955651
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:49827196 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACATTTATAACAATCGGGAGACTGA[C/T]TGGTAAAGGGAGAGGCTGCTGATCT
Phenotype
MIM: 606540
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MYO5B PubMed Links

Gene Details

Gene
MYO5B
Gene Name
myosin VB
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080467.2 Intron NP_001073936.1
Gene
SNHG22
Gene Name
small nucleolar RNA host gene 22
There are no transcripts associated with this gene.

View Full Product Details