Product Details

SNP ID
rs9999356
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.4:367130 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGGATAGGAAAGAGCAGAAATCTT[C/T]TTTTTTTGAGATCTGGAACAGGACA
Phenotype
MIM: 194648
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
ZNF141 PubMed Links

Gene Details

Gene
ZNF141
Gene Name
zinc finger protein 141
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003441.2 Intron NP_003432.1
XM_011513562.2 Intron XP_011511864.1
XM_011513563.2 Intron XP_011511865.1
XM_017008591.1 Intron XP_016864080.1

View Full Product Details