Product Details

SNP ID
rs10241898
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:2412093 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAGTTACTTTACTTCAAAGGCTAA[A/G]TTTGGGCTCAGTTTTCATTCAGGTC
Phenotype
MIM: 610129
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CHST12 PubMed Links

Gene Details

Gene
CHST12
Gene Name
carbohydrate (chondroitin 4) sulfotransferase 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243794.1 Intron NP_001230723.1
NM_001243795.1 Intron NP_001230724.1
NM_018641.4 Intron NP_061111.1
XM_011515443.2 Intron XP_011513745.1
XM_011515444.2 Intron XP_011513746.1

View Full Product Details