Product Details
- SNP ID
-
rs9309736
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:67244535 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TAAATGAATCCTTGCTTATTGAATT[C/G]CTCACTTTCTGCTACTAGCACCATC
- Phenotype
-
MIM: 603922
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
SUCLG2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs79164427] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SUCLG2
- Gene Name
- succinate-CoA ligase GDP-forming beta subunit
View Full Product Details