Product Details
- SNP ID
-
rs7417850
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
1
- Location
-
Chr.1:198530738 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGTGACAACTACATATATATTATTC[A/C]CAGAGATTCTCTGTGAGCAGAATAA
- Phenotype
-
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
ATP6V1G3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs77181329] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ATP6V1G3
- Gene Name
- ATPase H+ transporting V1 subunit G3
View Full Product Details