Product Details

SNP ID
rs10264956
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.7:86882911 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGAATCTTCTCCAAAGCCAACTTCC[C/T]CCAGAAGGTTTTCCTCACCTTGTCA
Phenotype
MIM: 614048
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
KIAA1324L PubMed Links

Gene Details

Gene
KIAA1324L
Gene Name
KIAA1324 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142749.2 Intron NP_001136221.1
NM_001291990.1 Intron NP_001278919.1
NM_001291991.1 Intron NP_001278920.1
NM_152748.3 Intron NP_689961.3
XM_006715894.3 Intron XP_006715957.1
XM_011515918.2 Intron XP_011514220.1
XM_011515921.1 Intron XP_011514223.1
XM_017011843.1 Intron XP_016867332.1
XM_017011844.1 Intron XP_016867333.1

View Full Product Details