Product Details

SNP ID
rs10213286
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:4860053 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCGCGGACGAGGAGGGGGCCAAG[C/T]CCAAAGTGTCCCCTTCGCTCCTGCC
Phenotype
MIM: 142983
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LINC01396 PubMed Links

Gene Details

Gene
LINC01396
Gene Name
long intergenic non-protein coding RNA 1396
There are no transcripts associated with this gene.

Gene
MSX1
Gene Name
msh homeobox 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002448.3 389 Missense Mutation CCC,TCC P52S NP_002439.2

View Full Product Details