Product Details

SNP ID
rs10422259
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:50662163 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGCCACGTCGAACTTGGTCCAGAA[C/T]CCCAGAGGTTTAGCGCCAAACGGCT
Phenotype
MIM: 604999 MIM: 600327
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C19orf81 PubMed Links

Gene Details

Gene
C19orf81
Gene Name
chromosome 19 open reading frame 81
There are no transcripts associated with this gene.

Gene
SHANK1
Gene Name
SH3 and multiple ankyrin repeat domains 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016148.3 6307 Silent Mutation GGA,GGG G2096G NP_057232.2
XM_006723233.3 6307 Silent Mutation GGA,GGG G2096G XP_006723296.1
XM_011527013.2 6307 Silent Mutation GGA,GGG G2104G XP_011525315.1
XM_011527014.2 6307 Silent Mutation GGA,GGG G2087G XP_011525316.1
Gene
SYT3
Gene Name
synaptotagmin 3
There are no transcripts associated with this gene.

View Full Product Details