Product Details

SNP ID
rs4855675
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.3:108912490 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCTCTCTTTCCTGATGCTTTTGTTC[A/G]TAATATAAAAATAAAACTAATTTGT
Phenotype
MIM: 605128
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
GUCA1C PubMed Links
Additional Information
For this assay, SNP(s) [rs139505400] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
GUCA1C
Gene Name
guanylate cyclase activator 1C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005459.3 Intron NP_005450.3
XM_005247910.3 Intron XP_005247967.1
XM_011513334.2 Intron XP_011511636.1

View Full Product Details