Product Details

SNP ID
rs10245273
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:158633269 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTATGGTCTATGTTCTGAGACATC[C/T]CTCCAAATTGACCTTTCTGGAATTT
Phenotype
MIM: 608532
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NCAPG2 PubMed Links

Gene Details

Gene
NCAPG2
Gene Name
non-SMC condensin II complex subunit G2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001281932.1 Intron NP_001268861.1
NM_001281933.1 Intron NP_001268862.1
NM_017760.6 Intron NP_060230.5
XM_005249547.2 Intron XP_005249604.1
XM_011516356.2 Intron XP_011514658.1
XM_011516357.2 Intron XP_011514659.1
XM_011516358.2 Intron XP_011514660.1
XM_011516362.1 Intron XP_011514664.1
XM_017012374.1 Intron XP_016867863.1
XM_017012375.1 Intron XP_016867864.1
XM_017012376.1 Intron XP_016867865.1
XM_017012377.1 Intron XP_016867866.1
XM_017012378.1 Intron XP_016867867.1
Gene
THAP5P1
Gene Name
THAP domain containing 5 pseudogene 1
There are no transcripts associated with this gene.

View Full Product Details