Product Details

SNP ID
rs10162574
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:105675090 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGCTGACACAGTTAGCTAAGAAAA[G/T]CCTGAAAAAAACAAAGTCTACAGGT
Phenotype
MIM: 146910 MIM: 147070 MIM: 147010
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ELK2AP PubMed Links

Gene Details

Gene
ELK2AP
Gene Name
ELK2A, member of ETS oncogene family, pseudogene
There are no transcripts associated with this gene.

Gene
IGH
Gene Name
immunoglobulin heavy locus
There are no transcripts associated with this gene.

View Full Product Details