Product Details

SNP ID
rs6629764
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:23909702 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTGGGAGCTGCATCCAAATTGGAGC[A/G]TTACTGCATCCAAACTCGTTGCTTT
Phenotype
MIM: 300753
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
APOO PubMed Links

Gene Details

Gene
APOO
Gene Name
apolipoprotein O
There are no transcripts associated with this gene.

Gene
CXorf58
Gene Name
chromosome X open reading frame 58
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001169574.1 Intron NP_001163045.1
NM_152761.2 Intron NP_689974.2
XM_011545473.2 Intron XP_011543775.1
XM_011545474.2 Intron XP_011543776.1

View Full Product Details