Product Details

SNP ID
rs9429201
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:46868820 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACAATTAATGTGTACTATTTTTCCA[C/T]CAACAACAGGCAAGCTAAGAGCCAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CYP4Z2P PubMed Links

Gene Details

Gene
CYP4Z2P
Gene Name
cytochrome P450 family 4 subfamily Z member 2, pseudogene
There are no transcripts associated with this gene.

View Full Product Details