Product Details

SNP ID
rs6002324
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:41310500 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGTCAGCACAAGGCACGAAAGCAA[A/G]CAGAGCAGCACTGGAAACTCTCTTG
Phenotype
MIM: 602362
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RANGAP1 PubMed Links

Gene Details

Gene
RANGAP1
Gene Name
Ran GTPase activating protein 1
There are no transcripts associated with this gene.

Gene
ZC3H7B
Gene Name
zinc finger CCCH-type containing 7B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017590.5 Intron NP_060060.3
XM_011530016.2 Intron XP_011528318.1

View Full Product Details