Product Details

SNP ID
rs6002332
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:41322941 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCTGCTGTGCCCAGCGTACTCCCTT[A/G]CCTTTTCTCACCCATGTCACCGCAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZC3H7B PubMed Links

Gene Details

Gene
ZC3H7B
Gene Name
zinc finger CCCH-type containing 7B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017590.5 Intron NP_060060.3
XM_011530016.2 Intron XP_011528318.1

View Full Product Details