Product Details

SNP ID
rs5943161
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.X:111515906 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCACTCTGAGAAACACTGCCCCAA[A/G]GGTTCCAAAACACTAGGGTTTCAGG
Phenotype
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
LINC00890 PubMed Links

Gene Details

Gene
LINC00890
Gene Name
long intergenic non-protein coding RNA 890
There are no transcripts associated with this gene.

View Full Product Details