Product Details

SNP ID
hCV30471866
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:119876660 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCAAAAAAAAAAAAAGAAGAAGAA[C/G]GATGGGTAATAATGGAATTTACCTC
Phenotype
MIM: 612974
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DEPTOR PubMed Links

Gene Details

Gene
DEPTOR
Gene Name
DEP domain containing MTOR-interacting protein
There are no transcripts associated with this gene.

View Full Product Details