Product Details

SNP ID
rs9904058
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:32275249 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCATGACTCGGCTTCAGTCCCCAGG[C/T]GGCGCGTCAGGCTGGGCCGGATGAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RHBDL3 PubMed Links

Gene Details

Gene
RHBDL3
Gene Name
rhomboid like 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138328.2 Intron NP_612201.1
XM_006721733.3 Intron XP_006721796.1
XM_006721734.3 Intron XP_006721797.1
XM_011524443.2 Intron XP_011522745.1
XM_017024272.1 Intron XP_016879761.1
XM_017024273.1 Intron XP_016879762.1
XM_017024274.1 Intron XP_016879763.1
XM_017024275.1 Intron XP_016879764.1
XM_017024276.1 Intron XP_016879765.1
XM_017024277.1 Intron XP_016879766.1
XM_017024278.1 Intron XP_016879767.1
XM_017024279.1 Intron XP_016879768.1
XM_017024280.1 Intron XP_016879769.1

View Full Product Details