Product Details
- SNP ID
-
rs9986248
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:140254743 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CCACTACCTCATATTCTTCCAGGTC[A/G]CTTCTGATACTCTCGCTCCAGCATT
- Phenotype
-
MIM: 604897
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PFDN1
PubMed Links
Gene Details
- Gene
- PFDN1
- Gene Name
- prefoldin subunit 1
View Full Product Details