Product Details

SNP ID
rs5767172
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.22:48496502 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAAGGTCACGTGGATGTGGAAACCC[G/T]TCTATACAATGATGTCAACCCTGCA
Phenotype
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
FAM19A5 PubMed Links

Gene Details

Gene
FAM19A5
Gene Name
family with sequence similarity 19 member A5, C-C motif chemokine like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001082967.2 Intron NP_001076436.1
NM_015381.6 Intron NP_056196.2

View Full Product Details