Product Details

SNP ID
rs9748330
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:14773631 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTAGTATCCTCCAAGTGGAGTTTAG[A/G]TTAAAATAATTTTAATCTCAACTCA
Phenotype
MIM: 616565
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ANKRD30B PubMed Links

Gene Details

Gene
ANKRD30B
Gene Name
ankyrin repeat domain 30B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145029.1 Intron NP_001138501.1
XM_006722325.3 Intron XP_006722388.1
XM_011525662.2 Intron XP_011523964.1
XM_011525664.2 Intron XP_011523966.1
XM_011525665.2 Intron XP_011523967.1
XM_011525666.2 Intron XP_011523968.1

View Full Product Details