Product Details

SNP ID
rs6504681
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.17:50736227 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTGGAATTTTGATTTACATGGTTTG[A/G]TTTGTAGTGTTATAAAATTAAATTT
Phenotype
MIM: 609434
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
LUC7L3 PubMed Links

Gene Details

Gene
LUC7L3
Gene Name
LUC7 like 3 pre-mRNA splicing factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006107.3 Intron NP_006098.2
NM_016424.4 Intron NP_057508.2
XM_005257448.1 Intron XP_005257505.1
XM_005257449.2 Intron XP_005257506.1
XM_005257452.1 Intron XP_005257509.1
XM_006721943.2 Intron XP_006722006.1

View Full Product Details