Product Details

SNP ID
rs10402702
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:44154123 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCAATGAGTAATAAAGCAGCCCTCA[C/T]TGGTGAGCTGACATTGAGCAGAGAC
Phenotype
MIM: 604750
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF234 PubMed Links

Gene Details

Gene
ZNF234
Gene Name
zinc finger protein 234
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144824.1 Intron NP_001138296.1
NM_006630.2 Intron NP_006621.1
XM_006722974.3 Intron XP_006723037.1
XM_017026149.1 Intron XP_016881638.1
XM_017026150.1 Intron XP_016881639.1
XM_017026151.1 Intron XP_016881640.1
XM_017026152.1 Intron XP_016881641.1
XM_017026153.1 Intron XP_016881642.1

View Full Product Details