Product Details

SNP ID
rs6610661
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:37783649 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTCCTCTAATTTTCAAAGGCCATCA[A/G]GCAAAATGCCCTTTTTTAGGTAAAA
Phenotype
MIM: 300481
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CYBB PubMed Links

Gene Details

Gene
CYBB
Gene Name
cytochrome b-245 beta chain
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000397.3 Intron NP_000388.2

View Full Product Details