Product Details

SNP ID
rs8009001
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:58402095 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGTTATATTAACTACAGTAGGATTT[A/C]TGCTATATGTGGCTGGGTAAAATTT
Phenotype
MIM: 607384
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
TIMM9 PubMed Links

Gene Details

Gene
TIMM9
Gene Name
translocase of inner mitochondrial membrane 9
There are no transcripts associated with this gene.

Gene
TOMM20L
Gene Name
translocase of outer mitochondrial membrane 20 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207377.2 Intron NP_997260.1
XM_011536742.2 Intron XP_011535044.1
XM_011536743.2 Intron XP_011535045.1
XM_011536744.2 Intron XP_011535046.1

View Full Product Details