Product Details
- SNP ID
-
rs5944882
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:101883820 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GTGTCGTTTTCTCTTCTGATGTTTC[C/T]GTTTATGACCTGCTTGATGGTCAGC
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZMAT1
PubMed Links
Gene Details
- Gene
- ZMAT1
- Gene Name
- zinc finger matrin-type 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001011657.3 |
1974 |
Missense Mutation |
CAG,CGG |
Q536R |
NP_001011657.2 |
NM_001282400.1 |
1974 |
Missense Mutation |
CAG,CGG |
Q365R |
NP_001269329.1 |
NM_001282401.1 |
1974 |
Missense Mutation |
CAG,CGG |
Q365R |
NP_001269330.1 |
XM_005262212.4 |
1974 |
Missense Mutation |
CAG,CGG |
Q616R |
XP_005262269.2 |
XM_005262213.4 |
1974 |
Missense Mutation |
CAG,CGG |
Q615R |
XP_005262270.2 |
XM_005262214.3 |
1974 |
Missense Mutation |
CAG,CGG |
Q593R |
XP_005262271.2 |
XM_005262216.4 |
1974 |
Missense Mutation |
CAG,CGG |
Q496R |
XP_005262273.2 |
XM_006724711.3 |
1974 |
Missense Mutation |
CAG,CGG |
Q365R |
XP_006724774.1 |
XM_017029903.1 |
1974 |
Missense Mutation |
CAG,CGG |
Q495R |
XP_016885392.1 |
XM_017029904.1 |
1974 |
Missense Mutation |
CAG,CGG |
Q365R |
XP_016885393.1 |
View Full Product Details