Product Details
- SNP ID
-
rs10165091
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:15591945 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGGACGGGGTGAAGATGGCGGCCTT[C/G]TCCGGTGCGTTTGTGGAAACTCTGG
- Phenotype
-
MIM: 601257
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
DDX1
PubMed Links
Gene Details
- Gene
- DDX1
- Gene Name
- DEAD/H-box helicase 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_004939.2 |
325 |
Silent Mutation |
TTC,TTG |
F4L |
NP_004930.1 |
View Full Product Details