Product Details

SNP ID
rs10119722
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:18927308 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCGTCGACTGCCTGGTTGCCAAGG[A/G]TGCTTTTTCTCCCTCCTCTCCCTGG
Phenotype
MIM: 616292
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SAXO1 PubMed Links

Gene Details

Gene
SAXO1
Gene Name
stabilizer of axonemal microtubules 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001287049.1 Intron NP_001273978.1
NM_001287050.1 Intron NP_001273979.1
NM_153707.3 Intron NP_714918.2
XM_011517744.1 Intron XP_011516046.1
XM_011517745.1 Intron XP_011516047.1
XM_011517746.2 Intron XP_011516048.1
XM_011517747.2 Intron XP_011516049.1
XM_017014314.1 Intron XP_016869803.1
XM_017014315.1 Intron XP_016869804.1

View Full Product Details