Product Details

SNP ID
rs6607452
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:74083702 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAAAACTAATAAGCAAGTTTAACAA[A/G]GTCACAGAAACCCATGATTAGCACA
Phenotype
MIM: 300936
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FTX PubMed Links

Gene Details

Gene
FTX
Gene Name
FTX transcript, XIST regulator (non-protein coding)
There are no transcripts associated with this gene.

View Full Product Details