Product Details

SNP ID
rs10418512
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:54713243 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGCAAGGAGGTAAATACATGAGA[A/G]AACAATGCACCCTTCAGAGTGCCAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LILRP2 PubMed Links

Gene Details

Gene
LILRP2
Gene Name
leukocyte immunoglobulin-like receptor pseudogene 2
There are no transcripts associated with this gene.

View Full Product Details