Product Details
- SNP ID
-
rs5759198
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:43155402 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GCTTGGAACCTTCTCCTGCCTCTAC[A/G]TCCTGCGCCCTCTACCCTGCCCCGA
- Phenotype
-
MIM: 109610
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TSPO
PubMed Links
Gene Details
- Gene
- TSPO
- Gene Name
- translocator protein
View Full Product Details